Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD23/Fc epsilon RII Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1231
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1231 500 μg
MAB1231SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1231 Supplier R&D Systems Supplier No. MAB1231
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD23/Fc epsilon RII Monoclonal specifically detects CD23/Fc epsilon RII in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CD23/Fc epsilon RII
Applications ELISA
Classification Monoclonal
Clone 138633
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P06734
Gene Alias BLAST-2, CD23, CD23A, CD23CD23 antigen, CLEC4J, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc epsilon RII, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, FCER2, Fcer2a, FceRII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Ly-42, Lymphocyte IgE receptor
Gene Symbols FCER2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD23 Met150-Ser321 Accession # P06734
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2208
Test Specificity Detects human CD23 in ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.