Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD24 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems FAB52471B100
This item is not returnable.
View return policy
Description
CD24 Polyclonal specifically detects CD24 in Human samples. It is validated for Flow Cytometry.Specifications
CD24 | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
NP_037362 | |
Sheep | |
Protein A or G purified | |
RUO | |
100133941 | |
Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG |
Flow Cytometry | |
Biotin | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
CD 24, CD24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24A, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen | |
Chinese hamster ovary cell line CHO-derived recombinant human CD24, Ser27-Gly59, Accession # NP_037362 | |
100 Tests | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction