Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD25/IL-2R alpha mFluor™ Violet 610-Conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. RD10000238
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10000238 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RD10000238 Supplier R&D Systems Supplier No. FAB1020MFV610100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD25/IL-2R alpha Monoclonal antibody specifically detects CD25/IL-2R alpha in Human samples. It is validated for nan

Specifications

Antigen CD25/IL-2R alpha
Applications Flow Cytometry
Classification Monoclonal
Clone 24212
Conjugate Unconjugated
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P01589
Gene Alias CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD25/IL-2 R alpha , Glu22-Cys213, Accession # P01589
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3559
Target Species Human
Content And Storage Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.