Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD25/IL-2R alpha PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
$139.40 - $303.50
Specifications
Antigen | CD25/IL-2R alpha |
---|---|
Clone | 24212 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1020P025
![]() |
R&D Systems
FAB1020P025 |
25 Tests |
Each for $139.40
|
|
|||||
FAB1020P100
![]() |
R&D Systems
FAB1020P100 |
100 Tests |
Each for $303.50
|
|
|||||
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Human samples. It is validated for Flow Cytometry.Specifications
CD25/IL-2R alpha | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
IL2RA | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human CD25/IL-2 R alpha in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-2 R gamma or rhIL-15 R alpha is observed. |
24212 | |
Flow Cytometry | |
R-PE | |
Mouse | |
Human | |
P01589 | |
3559 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD25/IL-2 R alpha Glu22-Cys213 Accession # P01589 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title