Learn More
Human CD25/IL-2R alpha (Research Grade Daclizumab Biosimilar) Alexa Fluor 405-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CD25/IL-2R alpha |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | Hu102 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Flow Cytometry 0.25-1 μg/10^6 cells |
| Formulation | Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide. |
| Gene Alias | CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR |
| Host Species | Human |
| Immunogen | Human CD25/IL-2 R alpha |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.