Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD28 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. RD10000194
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10000194 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RD10000194 Supplier R&D Systems Supplier No. FAB3422V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CD28
Applications Flow Cytometry
Classification Monoclonal
Clone 2946A
Conjugate Alexa Fluor 405
Dilution Flow Cytometry
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P10747
Gene Alias CD28, CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Host Species Rabbit
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD28, Asn19-Pro152, Accession # P10747
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 940
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.