Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD28 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | CD28 |
---|---|
Clone | 987701 |
Dilution | Immunohistochemistry 5-25 μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3421SP
![]() |
R&D Systems
MAB3421SP |
25 μg |
Each for $200.50
|
|
|||||
MAB3421100
![]() |
R&D Systems
MAB3421100 |
100 μg |
Each for $502.50
|
|
|||||
Description
CD28 Monoclonal specifically detects CD28 in Human samples. It is validated for Immunohistochemistry.Specifications
CD28 | |
Immunohistochemistry 5-25 μg/mL | |
Monoclonal | |
Lyophilized | |
Human | |
P10747 | |
940.0 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
987701 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD28, CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
Synthetic peptide containing human CD28, Accession No. P10747 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CD28 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title