Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD3 epsilon Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$221.50 - $456.00
Specifications
| Antigen | CD3 epsilon |
|---|---|
| Clone | UCHT1 |
| Dilution | Flow Cytometry 5 uL/10^6 cells, Immunocytochemistry 8-25 ug/mL |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB100T025
![]() |
R&D Systems
FAB100T025 |
25 Tests |
Each for $221.50
|
|
|||||
FAB100T100
![]() |
R&D Systems
FAB100T100 |
100 Tests |
Each for $456.00
|
|
|||||
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
| CD3 epsilon | |
| Flow Cytometry 5 uL/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 916 | |
| Human thymocytes (1) | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| UCHT1 | |
| Flow Cytometry, Immunocytochemistry | |
| Alexa Fluor 594 | |
| Mouse | |
| Human | |
| CD3e, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| CD3E | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Recognizes the epsilon-chain of the CD3/T cell antigen receptor complex (McMichael, A.J. et al. (1987) Leucocyte Typing III: White Cell Differentiation Antigens, Oxford University Press, New York; Knapp, W. et al. (1989) Leucocyte Typing IV: White Cell Differentiation Antigens, Oxford University Press, New York; Schlossman, S. et al. (1995) Leucocyte Typing V: White Cell Differentiation Antigens, Oxford University Press, New York). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title