Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD30 Ligand/TNFSF8 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10281S10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10281S10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10281S10 Supplier R&D Systems Supplier No. FAB10281S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD30 Ligand/TNFSF8 Monoclonal specifically detects CD30 Ligand/TNFSF8 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD30 Ligand/TNFSF8
Applications Flow Cytometry
Classification Monoclonal
Clone 116621
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P32971
Gene Alias CD153, CD153 antigen, CD30 antigen ligand, CD30L, CD30-L, CD30LCD30 ligand, CD30LGMGC138144, TNFSF8, tumor necrosis factor (ligand) superfamily, member 8, tumor necrosis factor ligand superfamily member 8
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived human CD30 Ligand/TNFSF8., Gln63-Asp234, Accession # P32971
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 944
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.