Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD30 Ligand/TNFSF8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$208.00 - $473.00
Specifications
| Antigen | CD30 Ligand/TNFSF8 |
|---|---|
| Clone | 116632 |
| Dilution | Flow Cytometry 0.25 ug/106 cells, Immunohistochemistry 5-25 ug/mL, Neutralization 0.3-3 ug/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Neutralization |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB7741SP
![]() |
R&D Systems
MAB7741SP |
25 μg |
Each for $208.00
|
|
|||||
MAB7741100
![]() |
R&D Systems
MAB7741100 |
100 μg |
Each for $473.00
|
|
|||||
Description
CD30 Ligand/TNFSF8 Monoclonal antibody specifically detects CD30 Ligand/TNFSF8 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, NeutralizationSpecifications
| CD30 Ligand/TNFSF8 | |
| Flow Cytometry 0.25 ug/106 cells, Immunohistochemistry 5-25 ug/mL, Neutralization 0.3-3 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD153, CD153 antigen, CD30 antigen ligand, CD30L, CD30-L, CD30LCD30 ligand, CD30LGMGC138144, TNFSF8, tumor necrosis factor (ligand) superfamily, member 8, tumor necrosis factor ligand superfamily member 8 | |
| Mouse myeloma cell line NS0-derived human CD30 Ligand/TNFSF8., Gln63-Asp234, Accession # P32971 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 116632 | |
| Flow Cytometry, Immunohistochemistry, Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| P32971 | |
| 944 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title