Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD30/TNFRSF8 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF229
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF229 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF229 Supplier R&D Systems Supplier No. BAF229
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CD30/TNFRSF8 Polyclonal specifically detects CD30/TNFRSF8 in Human samples. It is validated for Western Blot.

Specifications

Antigen CD30/TNFRSF8
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P28908
Gene Alias CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, TNFRSF8, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8
Gene Symbols TNFRSF8
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD30/TNFRSF8 Phe19-Lys379 Accession # P28908
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 943
Test Specificity Detects human CD30/TNFRSF8 in Western blots. In Western blots, less then 1% cross-reactivity with recombinant mouse CD30 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.