Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD300e/LMIR6 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2705R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2705R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2705R100 Supplier R&D Systems Supplier No. FAB2705R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD300e/LMIR6 Monoclonal specifically detects CD300e/LMIR6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD300e/LMIR6
Applications Flow Cytometry
Classification Monoclonal
Clone 233810
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias CD300 antigen like family member E, CD300 antigen-like family member E, CD300e, CD300e antigenPIgR-2, CD300e molecule, CD300LE, CD300LEPIgR2, CLM2, CLM-2, CLM2CMRF35-A5, CMRF35A5, CMRF35-like molecule 2, Immune receptor expressed on myeloid cells 2, IREM-2, IREM2IREM-2, LMIR6, Poly-Ig receptor 2, Polymeric immunoglobulin receptor 2
Gene Symbols CD300E
Host Species Rat
Immunogen Y3 rat myeloid cell line transfected with human CD300e/LMIR6
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342510
Test Specificity Detects human CD300e/LMIR6 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human LMIR1, 2, 3, 4, or 5 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.