Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD300f/LMIR3 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2774S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2774S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2774S100 Supplier R&D Systems Supplier No. FAB2774S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD300f/LMIR3/CD300LF Monoclonal specifically detects CD300f/LMIR3/CD300LF in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD300f/LMIR3/CD300LF
Applications Flow Cytometry
Classification Monoclonal
Clone 234903
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias CMRF35-like molecule 1, CD300 molecule-like family member f, CD300f, CD300LF, CLIM1, CLM1, CLM-1, DIgR2, IgSF13, IREM1, IREM-1, LMIR3, NKIR, Pigr3
Gene Symbols CD300LF
Host Species Rat
Immunogen Y3 rat myeloid cell line transfected with human CD300f/LMIR3
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146722
Test Specificity Detects human CD300f/LMIR3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human LMIR1, 2, 4, 5, 6, or recombinant mouse LMIR3 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.