Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD300f/LMIR3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2774SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2774SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2774SP Supplier R&D Systems Supplier No. MAB2774SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

CD300f/LMIR3/CD300LF Monoclonal specifically detects CD300f/LMIR3/CD300LF in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen CD300f/LMIR3/CD300LF
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 234903
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias CMRF35-like molecule 1, CD300 molecule-like family member f, CD300f, CD300LF, CLIM1, CLM1, CLM-1, DIgR2, IgSF13, IREM1, IREM-1, LMIR3, NKIR, Pigr3
Gene Symbols CD300LF
Host Species Rat
Immunogen Y3 rat myeloid cell line transfected with human CD300f/LMIR3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146722
Test Specificity Detects human CD300f/LMIR3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human LMIR1, 2, 4, 5, 6, or recombinant mouse LMIR3 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.