Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD300g/Nepmucin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5328
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5328 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5328 Supplier R&D Systems Supplier No. AF5328
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

CD300LG/Nepmucin Polyclonal specifically detects CD300LG/Nepmucin in Human samples. It is validated for Western Blot.

Specifications

Antigen CD300LG/Nepmucin
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q6UXG3
Gene Alias CD300 antigen like family member G, CD300 antigen-like family member G, CD300 molecule like family member g, CD300 molecule-like family member g, CD300g antigen, CD300LG, CLM9, CLM-9, CLM9Trem4, CMRF35-like molecule 9, NEPMUCIN, TREM4, TREM-4, Triggering receptor expressed on myeloid cells 4
Gene Symbols CD300LG
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD300g/Nepmucin Leu19-Leu249 Accession # Q6UXG3
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146894
Test Specificity Detects human CD300g/Nepmucin in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse CD300g/Nepmucin is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.