Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD302/CLEC13A Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6367G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6367G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6367G100 Supplier R&D Systems Supplier No. FAB6367G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD302/CLEC13A Monoclonal specifically detects CD302/CLEC13A in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD302/CLEC13A
Applications Flow Cytometry
Classification Monoclonal
Clone 771910
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q81X05
Gene Alias BIMLEC, CD302, CD302 antigenC-type lectin domain family 13, member A, CD302 molecule, CLEC13A, CLEC13AMGC22301, C-type lectin domain family 13 member A, DCL1, DCL-1, DCL1C-type lectin BIMLEC, KIAA0022FLJ43091, Type I transmembrane C-type lectin receptor DCL-1
Gene Symbols CD302
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD302/CLEC13A, Asp23-Asn167, Accession # Q81X05
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 9936
Test Specificity Detects human CD302/CLEC13A in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse CD302 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.