Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD302/CLEC13A Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6367R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6367R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6367R100 Supplier R&D Systems Supplier No. FAB6367R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD302/CLEC13A Monoclonal specifically detects CD302/CLEC13A in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD302/CLEC13A
Applications Flow Cytometry
Classification Monoclonal
Clone 771910
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q81X05
Gene Alias BIMLEC, CD302, CD302 antigenC-type lectin domain family 13, member A, CD302 molecule, CLEC13A, CLEC13AMGC22301, C-type lectin domain family 13 member A, DCL1, DCL-1, DCL1C-type lectin BIMLEC, KIAA0022FLJ43091, Type I transmembrane C-type lectin receptor DCL-1
Gene Symbols CD302
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD302/CLEC13A Asp23-Asn167 Accession # Q81X05
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9936
Test Specificity Detects human CD302/CLEC13A in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse CD302 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.