Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD302/CLEC13A Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6367
This item is not returnable.
View return policy
Description
CD302/CLEC13A Monoclonal specifically detects CD302/CLEC13A in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
CD302/CLEC13A | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Q81X05 | |
CD302 | |
Mouse myeloma cell line NS0-derived recombinant human CD302/CLEC13A Asp23-Asn167 Accession # Q81X05 | |
100 μg | |
Primary | |
Detects human CD302/CLEC13A in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse CD302 is observed. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
771910 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BIMLEC, CD302, CD302 antigenC-type lectin domain family 13, member A, CD302 molecule, CLEC13A, CLEC13AMGC22301, C-type lectin domain family 13 member A, DCL1, DCL-1, DCL1C-type lectin BIMLEC, KIAA0022FLJ43091, Type I transmembrane C-type lectin receptor DCL-1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9936 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction