Learn More
Human CD31/PECAM-1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CD31/PECAM-1 |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Dilution | Flow Cytometry 5μL/106 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. |
| Gene Alias | adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule |
| Host Species | Sheep |
| Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human CD31 extracellular domain |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.