Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD31/PECAM-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 25 publications
Supplier: R&D Systems AF806SP
This item is not returnable.
View return policy
Description
CD31/PECAM-1 Polyclonal specifically detects CD31/PECAM-1 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry, Knockout Validated.Specifications
CD31/PECAM-1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
PECAM1 | |
Chinese hamster ovary cell line CHO-derived recombinant human CD31 Extracellular domain | |
25 μg | |
Primary | |
Detects human CD31 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse CD31 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoassay | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-15 ug/mL, Knockout Validated 5-15 ug/mL | |
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
Sheep | |
Affinity Purified | |
RUO | |
5175 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction