Learn More
Human CD31/PECAM-1 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 4 publications
Supplier:  R&D Systems FAB3567A100
 View more versions of this product
Catalog No. FAB3567A100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human samples. It is validated for Flow Cytometry, Knockout Validated.
Specifications
Specifications
CD31/PECAM-1
Monoclonal
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
PECAM1
Activated HUVEC human umbilical vein endothelial cells
100 Tests
Primary
Detects human CD31/PECAM-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) E-Selectin, rhICAM-1, -2, -3, rhVCAM-1, or recombinant mouse VCAM-1 is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Flow Cytometry, Immunoassay
9G11
Flow Cytometry 10 μL/10^6 cells, Knockout Validated
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
5175
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.