Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human CD31/PECAM-1 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP191175 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP191175 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP191175 Supplier Novus Biologicals™ Supplier No. NBP191175

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human CD31/PECAM-1 ELISA Kit (Colorimetric) measures human CD31/PECAM-1 in cell culture supernatant, serum, plasma (edta, citrate, heparin), urine.

Specifications

Assay Range 0.47 - 30.0ng/mL
Assay Sensitivity 0.06ng/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 5175
Gene Symbol PECAM1
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sPECAM-1, HRP-Conjugate anti-human sPECAM-1 monoclonal antibody, Human sPECAM-1 Standard, 60ng/mL, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Angiogenesis, Cancer, CD Markers, Cellular Markers, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Signal
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Target CD31/PECAM-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.