Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD320/TCblR/8D6A Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | CD320/TCblR/8D6A |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
CD320/TCblR/8D6A Polyclonal specifically detects CD320/TCblR/8D6A in Human samples. It is validated for Western Blot.Specifications
CD320/TCblR/8D6A | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
8D6 antigen, 8D6A, 8D6AFDC-SM-8D6, CD320, CD320 antigenFDC-signaling molecule 8D6,8D6TCBLR, CD320 molecule, TCblR, Transcobalamin receptor | |
CD320 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9NPF0 | |
51293 | |
Mouse myeloma cell line NS0-derived recombinant human CD320/TCblR/8D6A Leu31-Val231 Accession # Q9NPF0 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human CD320/TCblR/8D6A in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title