Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD34 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB11441V100UG
This item is not returnable.
View return policy
Description
CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Flow CytometrySpecifications
CD34 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD34, CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule, HPCA1 | |
Chinese Hamster Ovary cell line, CHO-derived human CD34, Ser32-Thr290, Accession # P28906 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2936H | |
Flow Cytometry | |
P28906 | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
947 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction