Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD34 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7227T100UG
This item is not returnable.
View return policy
Description
CD34 Monoclonal specifically detects CD34 in Human samples. It is validated for Flow Cytometry.Specifications
CD34 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human CD34 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
QBEnd10 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
CD34, CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule, HPCA1 | |
Human endothelial vesicles | |
100 μg | |
947 | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction