Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD36/SR-B3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $514.50
Specifications
Antigen | CD36/SR-B3 |
---|---|
Clone | 1283D |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB19554SP
![]() |
R&D Systems
MAB19554SP |
25 μg |
Each for $214.00
|
|
|||||
MAB19554100
![]() |
R&D Systems
MAB19554100 |
100 μg |
Each for $514.50
|
|
|||||
Description
CD36/SR-B3 Monoclonal specifically detects CD36/SR-B3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
CD36/SR-B3 | |
Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
BDPLT10, CD_antigen: CD36, CD36, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor), CD36 molecule (thrombospondin receptor), CHDS7, cluster determinant 36, Collagen R, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, PAS IV, PAS-4, PAS-4 protein, PASIV, Platelet collagen receptor, platelet glycoprotein 4, SCARB3, scavenger receptor class B, member 3, SRB3, SR-B3, Thrombospondin R, Thrombospondin receptor | |
Human CD36/SR-B3 synthetic peptide Accession # P16671 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CD36/SR-B3 in direct ELISAs and Western blots. |
1283D | |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P16671 | |
948 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title