Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD39L1/ENTPD2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6087SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6087SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6087SP Supplier R&D Systems Supplier No. AF6087SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

CD39L1/ENTPD2 Polyclonal specifically detects CD39L1/ENTPD2 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen CD39L1/ENTPD2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Y5L3
Gene Alias CD39 antigen-like 1, CD39L1, CD39L1NTPDase 2, CD39-like-1, EC 3.6.1, EC 3.6.1.-, EC 3.6.1.5, Ecto-ATP diphosphohydrolase 2, Ecto-ATPase, Ecto-ATPase 2, Ecto-ATPDase 2, ectonucleoside triphosphate diphosphohydrolase 2, ENTPD2, NTPD2, NTPDase 2, NTPDase-2
Gene Symbols ENTPD2
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human CD39L1/ENTPD2 Thr29-Asp460 Accession # Q9Y5L3
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 954
Test Specificity Detects human CD39L1 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse CD39L1 and less than 1% cross-reactivity with recombinant human (rh) CD39L2, rhCD39L3, and rhCD39L4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.