Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD39L3/ENTPD3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4400
This item is not returnable.
View return policy
Description
CD39L3/ENTPD3 Polyclonal specifically detects CD39L3/ENTPD3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| CD39L3/ENTPD3 | |
| Polyclonal | |
| Unconjugated | |
| O75355 | |
| ENTPD3 | |
| Mouse myeloma cell line NS0-derived recombinant human CD39L3/ENTPD3 Gln44-Pro485 Accession # O75355 | |
| 100 μg | |
| Primary | |
| Detects human CD39L3/ENTPD3 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse CD39L3 is observed, and less than 2% cross-reactivity with recombinant human (rh) CD39L1 and rhCD39 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| ATPDase, CD39 antigen-like 3, CD39L3, CD39L3EC 3.6.1.5, CD39-like 3, EC 3.6.1, Ecto-apyrase 3, Ecto-ATP diphosphohydrolase 3, Ecto-ATPase 3, Ecto-ATPDase 3, ectonucleoside triphosphate diphosphohydrolase 3, ENTPD3, FLJ93839, HB6, HB6NTPDase 3, NTPDase-3 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 956 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction