Learn More
Human CD4 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB3791N025
 View more versions of this product
Catalog No. FAB3791N025
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD4
Monoclonal
Alexa Fluor 700
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
920
Human
Purified
Flow Cytometry
11830
Flow Cytometry 5 uL/10^6 cells
P01730
CD4
Recombinant Human CD4. Extracellular Domain Accession # P01730
25 Tests
Primary
Detects human CD4 in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.