Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD4 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$141.13 - $271.82

Specifications

Antigen CD4
Clone 11830
Dilution Flow Cytometry 5 uL/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB3791S025
GREENER_CHOICE
View Documents
R&D Systems
FAB3791S025
25 Tests
Each for $141.13
Only null left
Add to Cart
 
FAB3791S100
GREENER_CHOICE
View Documents
R&D Systems
FAB3791S100
100 Tests
Each for $271.82
Only null left
Add to Cart
 
Description

Description

CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD4
Flow Cytometry 5 uL/10^6 cells
Monoclonal
Purified
RUO
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
CD4
IgG2a
Protein A or G purified from hybridoma culture supernatant
Detects human CD4 in direct ELISAs and Western blots. Does not cross-react with recombinant mouse CD4.
11830
Flow Cytometry
Alexa Fluor 750
Mouse
Human
P01730
920
Recombinant human CD4 Extracellular domain Accession # P01730
Primary
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.