Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
Supplier: R&D Systems MAB379500
This item is not returnable.
View return policy
Description
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
CD4 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD4 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD4 Extracellular domain | |
500 μg | |
Primary | |
Detects human CD4 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse CD4 is observed. | |
Human | |
Ascites |
Flow Cytometry, Immunocytochemistry, CyTOF | |
34930 | |
Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
920 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction