Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | CD4 |
---|---|
Clone | 1033236 |
Dilution | Flow Cytometry 0.25 ug/106 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB37913SP
![]() |
R&D Systems
MAB37913SP |
25 μg |
Each for $200.50
|
|
|||||
MAB37913100
![]() |
R&D Systems
MAB37913100 |
100 μg |
Each for $467.50
|
|
|||||
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow CytometrySpecifications
CD4 | |
Flow Cytometry 0.25 ug/106 cells | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
Chinese Hamster Ovary cell line CHO-derived human CD4, Lys26-Trp390, Accession # P01730.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1033236 | |
Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
P01730.1 | |
920 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title