Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD40/TNFRSF5 Antibody, R&D Systems™



Mouse Monoclonal Antibody has been used in 5 publications
Supplier: R&D Systems MAB6321100
This item is not returnable.
View return policy
Description
CD40/TNFRSF5 Monoclonal specifically detects CD40/TNFRSF5 in Human samples. It is validated for Flow Cytometry, Agonist Activity, Immunocytochemistry, CyTOF-ready.Specifications
| CD40/TNFRSF5 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Detects human CD40/TNFRSF5 in direct ELISAs and Western blots. In direct ELISAs, does not cross-react with recombinant human (rh) 4-1BB, rhCD27, rhCD30, recombinant mouse CD40, rhDR3, rhDR6, rhEDAR, rhFas, rhGITR, rhHVEM, rhLTR beta , rhNGF R, rhOPG, rhRANK, rhTAJ, rhTNF RI, or rhTNF RII. | |
| Human | |
| Supernatant |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 82111 | |
| Flow Cytometry 2.5 ug/10^6 cells, Agonist Activity, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| P25942 | |
| CD40 | |
| Mouse myeloma cell line NS0-derived recombinant human CD40/TNFRSF5 Glu21-Arg193 Accession # P25942 | |
| 100 μg | |
| Agonist Activity | |
| 958 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction