Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD40/TNFRSF5 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6321A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6321A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6321A Supplier R&D Systems Supplier No. FAB6321A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD40/TNFRSF5 Monoclonal specifically detects CD40/TNFRSF5 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD40/TNFRSF5
Applications Flow Cytometry
Classification Monoclonal
Clone 82111
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P25942
Gene Alias B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5
Gene Symbols CD40
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD40/TNFRSF5 Glu21-Arg193 Accession # P25942
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 958
Test Specificity Detects human CD40/TNFRSF5 in direct ELISAs and Western blots. In direct ELISAs, does not cross-react with recombinant human (rh) 4-1BB, rhCD27, rhCD30, recombinant mouse CD40, rhDR3, rhDR6, rhEDAR, rhFas, rhGITR, rhHVEM, rhLTR beta , rhNGF R, rhOPG, rhRANK, rhTAJ, rhTNF RI, or rhTNF RII.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.