Learn More
Human CD42b/GPIb alpha Alexa Fluor 647-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CD42b/GPIb alpha |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 486805 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | P07359 |
| Gene Alias | Antigen CD42b-alpha, BP1BA, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP1BA, GPIb alpha, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.