Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD42b/GPIb alpha APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB4067A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4067A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4067A Supplier R&D Systems Supplier No. FAB4067A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD42b/GPIb alpha Monoclonal specifically detects CD42b/GPIb alpha in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD42b/GPIb alpha
Applications Flow Cytometry
Classification Monoclonal
Clone 486805
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P07359.1
Gene Alias Antigen CD42b-alpha, BP1BA, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP1BA, GPIb alpha, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit
Gene Symbols GP1BA
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD42b/GPIb alpha His17-Leu505 Accession # P07359.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2811
Test Specificity Detects human CD42b/GPIb alpha in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.