Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD45 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1430T025
This item is not returnable.
View return policy
Description
CD45 Monoclonal specifically detects CD45 in Human samples. It is validated for Flow Cytometry.Specifications
CD45 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 5 uL/10^6 cells | |
B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5788 | |
Human | |
Purified |
Flow Cytometry | |
2D1 | |
Alexa Fluor 594 | |
Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
PTPRC | |
Human peripheral blood mononuclear cells | |
25 Tests | |
Primary | |
Detects human CD45. This antibody recognizes all isoforms of human CD45. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction