Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD45 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$208.00 - $460.50
Specifications
Antigen | CD45 |
---|---|
Clone | 1043759 |
Dilution | Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 5-25 ug/mL, Knockout Validated |
Applications | Western Blot, Immunohistochemistry, KnockDown |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB14303SP
![]() |
R&D Systems
MAB14303SP |
25 μg |
Each for $208.00
|
|
|||||
MAB14303100
![]() |
R&D Systems
MAB14303100 |
100 μg |
Each for $460.50
|
|
|||||
Description
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, KnockDownSpecifications
CD45 | |
Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 5-25 ug/mL, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
B220, CD_antigen: CD45, CD45, CD45 antigen, CD45R, EC 3.1.3.48, EC:3.1.3.48, GP180, LCA, L-CA, Leukocyte common antigen, LY5, protein tyrosine phosphatase, receptor type, c polypeptide, PTPRC, receptor-type tyrosine-protein phosphatase C, T200, T200 glycoprotein, T200 leukocyte common antigen | |
Mouse myeloma cell line NS0-derived human CD45 protein, Gln24-Lys575, Accession # P08575 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1043759 | |
Western Blot, Immunohistochemistry, KnockDown | |
Unconjugated | |
Mouse | |
Human | |
P08575 | |
5788 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title