Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD45 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
$162.90 - $374.00
Specifications
| Antigen | CD45 |
|---|---|
| Clone | 2D1 |
| Dilution | Flow Cytometry 10 μL/106 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1430A025
![]() |
R&D Systems
FAB1430A025 |
25 Tests |
Each for $162.90
|
|
|||||
FAB1430A100
![]() |
R&D Systems
FAB1430A100 |
100 Tests |
Each for $374.00
|
|
|||||
Description
CD45 Monoclonal specifically detects CD45 in Human samples. It is validated for Flow Cytometry.Specifications
| CD45 | |
| Flow Cytometry 10 μL/106 cells | |
| Monoclonal | |
| Supernatant | |
| Human | |
| B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| PTPRC | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human CD45. This antibody recognizes all isoforms of human CD45. |
| 2D1 | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 5788.0 | |
| Human peripheral blood mononuclear cells | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title