Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD45RA Antibody, R&D Systems™

Mouse Monoclonal Antibody
$208.00 - $474.50
Specifications
Antigen | CD45RA |
---|---|
Clone | 1069433 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL |
Applications | Flow Cytometry, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11444100
![]() |
R&D Systems
MAB11444100 |
100 μg |
Each for $474.50
|
|
|||||
MAB11444SP
![]() |
R&D Systems
MAB11444SP |
25 μg |
Each for $208.00
|
|
|||||
Description
CD45RA Monoclonal antibody specifically detects CD45RA in Human samples. It is validated for Flow Cytometry, ImmunohistochemistrySpecifications
CD45RA | |
Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Whole cells (naïve T cells), Accession # P08575 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1069433 | |
Flow Cytometry, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
P08575 | |
5788 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title