Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD45RB Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB11434N100UG
This item is not returnable.
View return policy
Description
CD45RB Monoclonal antibody specifically detects CD45RB in Human samples. It is validated for Flow CytometrySpecifications
CD45RB | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Chinese Hamster Ovary cell line, CHO-derived human CD45RB, Gln26-Lys463, Accession # P08575 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1068228 | |
Flow Cytometry | |
P08575 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5788 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction