Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD45RO Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10642T10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10642T10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10642T10 Supplier R&D Systems Supplier No. FAB10642T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD45RO Monoclonal antibody specifically detects CD45RO in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CD45RO
Applications Flow Cytometry
Classification Monoclonal
Clone UCHL1
Conjugate Alexa Fluor 594
Dilution Flow Cytometry
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Host Species Mouse
Immunogen Cultured T cells from an IL-2-dependent T-cell line (CA1) prepared from human peripheral blood activated with influenza virus.
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5788
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.