Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1636SP
This item is not returnable.
View return policy
Description
CD5 Monoclonal specifically detects CD5 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
CD5 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
P06127 | |
CD5 | |
Mouse myeloma cell line NS0-derived recombinant human CD5 Arg25-Asn371 Accession # P06127 | |
25 μg | |
Primary | |
Detects human CD5 in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
205919 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD5, CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
921 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction