Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD52 (Research Grade Alemtuzumab Biosimilar) Alexa Fluor 700-conjugated Antibody, R&D Systems™

Human Monoclonal Antibody
Supplier: R&D Systems FAB9889N100UG
This item is not returnable.
View return policy
Description
CD52 Monoclonal specifically detects CD52 in Human samples. It is validated for Flow Cytometry.Specifications
CD52 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
Human | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human CD52 based on Alemtuzumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Alemtuzumab. This product is for research use only. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG |
Flow Cytometry | |
Hu116 | |
Flow Cytometry 0.25-1μg/106 cells | |
Cambridge pathology 1 antigen, CAMPATH-1, CAMPATH-1 antigen, CD52, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, HEL-S-171mP, Human epididymis-specific protein 5 | |
Human CD52 | |
100 μg | |
1043 | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction