Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD52 (Research Grade Alemtuzumab Biosimilar) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9889100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9889100 100 μg
MAB9889SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9889100 Supplier R&D Systems Supplier No. MAB9889100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Monoclonal Antibody

CD52 Monoclonal specifically detects CD52 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen CD52
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone Hu116
Conjugate Unconjugated
Dilution Flow Cytometry 0.25μg/106 cells, CyTOF-ready
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Alias Cambridge pathology 1 antigen, CAMPATH-1, CAMPATH-1 antigen, CD52, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, HEL-S-171mP, Human epididymis-specific protein 5
Host Species Human
Immunogen Human CD52
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1043
Test Specificity Detects human CD52 based on Alemtuzumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Alemtuzumab. This product is for research use only.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.