Learn More
Human CD52 (Research Grade Alemtuzumab Biosimilar) Biotinylated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CD52 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | Hu116 |
| Conjugate | Biotin |
| Dilution | Flow Cytometry 10 μL/106 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. |
| Gene Alias | Cambridge pathology 1 antigen, CAMPATH-1, CAMPATH-1 antigen, CD52, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, HEL-S-171mP, Human epididymis-specific protein 5 |
| Host Species | Human |
| Immunogen | Human CD52 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.