Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD55/DAF Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2009SP
This item is not returnable.
View return policy
Description
CD55/DAF Monoclonal specifically detects CD55/DAF in Human samples. It is validated for Western Blot, Neutralization.Specifications
CD55/DAF | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Neutralization 0.5-2 ug/mL | |
P08174.4 | |
CD55 | |
NS0-derived recombinant human CD55/DAF Asp35-Ser353 Accession # P08174.4 | |
25 μg | |
Primary | |
Detects human CD55/DAF in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CD97 is observed. | |
Human | |
Purified |
Western Blot, Neutralization | |
278803 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD55, CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CR, CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, DAF, decay accelerating factor for complement, TC | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1604 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction