Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD58/LFA-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $667.00
Specifications
Antigen | CD58/LFA-3 |
---|---|
Clone | 248310 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Description
CD58/LFA-3 Monoclonal specifically detects CD58/LFA-3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
CD58/LFA-3 | |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
P19256 | |
965 | |
Mouse myeloma cell line NS0-derived recombinant human CD58/LFA-3 Phe29-Arg215 Accession # P19256 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CD58/LFA-3 in direct ELISAs and Western blots. |
248310 | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Ag3, CD58, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA3, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3 | |
CD58 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title