Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD5L Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $608.00
Specifications
Antigen | CD5L |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
CD5L Polyclonal specifically detects CD5L in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CD5L | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
AIM, API6, apoptosis inhibitor 6, CD5 antigen-like, CD5 antigen-like (scavenger receptor cysteine rich family), CD5 molecule-like, CD5L, CT-2, IgM-associated peptide, PRO229, Spalpha, SP-ALPHA | |
CD5L | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
O43866 | |
922 | |
Mouse myeloma cell line NS0-derived recombinant human CD5L, Ser20-Gly347, Accession # O43866 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human CD5L in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant mouse CD5L and less than 1% cross-reactivity with recombinant human CD5 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title