Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD63 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $479.50
Specifications
Antigen | CD63 |
---|---|
Clone | 2585J |
Dilution | Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 0.3-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB50482SP
![]() |
R&D Systems
MAB50482SP |
25 μg |
Each for $200.50
|
|
|||||
MAB50482100
![]() |
R&D Systems
MAB50482100 |
100 μg |
Each for $479.50
|
|
|||||
Description
CD63 Monoclonal specifically detects CD63 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
CD63 | |
Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 0.3-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, Lamp-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30, TSPAN30granulophysin | |
Chinese Hamster Ovary cell line CHO-derived human CD63, Ala103-Val203, Accession # P08962-1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2585J | |
Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF | |
Unconjugated | |
Rabbit | |
Human | |
P08962-1 | |
967 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title